siRNA Details
| Gene Symbol: Vmn1r159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166758.1 | NP_001160230.1 | 1 | 705 | ||
| Gene Symbol: Vmn1r113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166716.1 | NP_001160188.1 | 1 | 711 | ||

Vmn1r159
Genomic Map