siRNA Details
Gene Symbol: B020004J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033790.3 | NP_001028962.1 | 3 | 514 | ||
GenBank mRNA | AK139557.1 | - | 3 | 515 | ||
BC147376.1 | - | 2 | 410 | 9056473 | ||
BC147377.1 | - | 2 | 410 | 9056471 |
