siRNA Details
| Gene Symbol: Ncoa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019825.3 | NP_062799.2 | 10 | 5648 | ||
| GenBank mRNA | AF216186.1 | - | 9 | 5533 | ||
| AK129079.1 | - | 3 | 2807 | |||
| AK142566.1 | - | 4 | 3833 | |||
| BC083187.1 | - | 10 | 5648 | 30544739 | ||
| JF707635.1 | - | 9 | 5507 | |||

Ncoa6
Genomic Map