siRNA Details
| Gene Symbol: Clec1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204239.1 | NP_001191168.1 | 5 | 843 | ||
| NM_001204253.1 | NP_001191182.1 | 4 | 691 | |||
| NM_019985.3 | NP_064369.1 | 6 | 939 | |||
| GenBank mRNA | AF201457.1 | - | 6 | 650 | ||
| AK007925.1 | - | 6 | 919 | |||
| AK154961.1 | - | 194 | ||||
| BC064054.1 | - | 6 | 939 | 574821 | ||
| EF070191.1 | - | 5 | 533 | |||
| EF070192.1 | - | 4 | 378 | |||

Clec1b
Genomic Map