siRNA Details
| Gene Symbol: Hbb-bh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127686.1 | NP_001121158.1 | 1 | 175 | ||
| GenBank mRNA | BC147125.1 | - | 1 | 175 | 9056191 | |
| BC147126.1 | - | 1 | 175 | 9056176 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hbb-bh2| Gene Symbol: Hbb-bh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127686.1 | NP_001121158.1 | 1 | 175 | ||
| GenBank mRNA | BC147125.1 | - | 1 | 175 | 9056191 | |
| BC147126.1 | - | 1 | 175 | 9056176 | ||