siRNA Details
| Gene Symbol: Adh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288578.1 | NP_001275507.1 | 8 | 1095 | ||
| NM_007410.3 | NP_031436.2 | 8 | 1113 | |||
| GenBank mRNA | AK002790.1 | - | 8 | 1035 | ||
| AK076507.1 | - | 8 | 1042 | |||
| AK146949.1 | - | 8 | 1057 | |||
| AK159803.1 | - | 8 | 1093 | |||
| BC062879.1 | - | 8 | 1039 | 5691327 | ||
| BC090978.1 | - | 8 | 1036 | 30313517 | ||
| M84147.1 | - | 8 | 1063 | |||

Adh5
Genomic Map