siRNA Details
| Gene Symbol: Ostc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025509.3 | NP_079785.1 | 3 | 406 | ||
| GenBank mRNA | AK009237.1 | - | 3 | 381 | ||
| AK011085.1 | - | 3 | 358 | |||
| AK013011.1 | - | 3 | 365 | |||
| AK135332.1 | - | 3 | 362 | |||
| BC014799.1 | - | 3 | 381 | |||
| BC021935.1 | - | 3 | 380 | 5149713 | ||
| BY228396.1 | - | 3 | 408 | |||

Ostc
Genomic Map