siRNA Details
| Gene Symbol: Trpd52l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025741.2 | NP_080017.1 | 1 | 1704 | ||
| GenBank mRNA | AK016450.1 | - | 1 | 1704 | ||
| BC125314.1 | - | 1 | 1705 | 40129829 | ||
| BC125316.1 | - | 1 | 1705 | 40129831 | ||

Trpd52l3
Genomic Map