siRNA Details
| Gene Symbol: Gpr89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026229.4 | NP_080505.1 | 8 | 714 | ||
| GenBank mRNA | AK002937.1 | - | 8 | 733 | ||
| AK009734.1 | - | 7 | 593 | |||
| AK016787.1 | - | 8 | 714 | |||
| AK034931.1 | - | 282 | ||||
| BC010729.1 | - | 8 | 707 | 3708143 | ||
More Information
Set Membership: |
|

Gpr89
Genomic Map