siRNA Details
| Gene Symbol: Mesdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023403.3 | NP_075892.3 | 3 | 784 | ||
| GenBank mRNA | AK008735.1 | - | 3 | 501 | ||
| AK031949.1 | - | 3 | 527 | |||
| AK076760.1 | - | 3 | 524 | |||
| AK076773.1 | - | 3 | 519 | |||
| BC014742.1 | - | 3 | 505 | 4239248 | ||
| BC058612.1 | - | 3438 | ||||

Mesdc2
Genomic Map