siRNA Details
| Gene Symbol: Kansl1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122738.1 | NP_001116210.1 | 2 | 811 | ||
| NM_177645.4 | NP_808313.3 | 2 | 811 | |||
| GenBank mRNA | AK081413.1 | - | 3 | 1161 | ||
| AK138707.1 | - | 1 | 642 | |||
| AK145440.1 | - | 2 | 782 | |||
| AK170981.1 | - | 2 | 840 | |||
| AK172645.1 | - | 2 | 764 | |||
| AK220534.1 | - | 2 | 762 | |||
| BC141036.1 | - | 3 | 721 | 9055648 | ||

Kansl1l
Genomic Map