siRNA Details
| Gene Symbol: Hddc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027168.2 | NP_081444.1 | 5 | 498 | ||
| GenBank mRNA | AK009957.1 | - | 5 | 439 | ||
| BC104360.1 | - | 5 | 498 | 40049707 | ||
| BC104361.1 | - | 5 | 498 | 40049709 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hddc2| Gene Symbol: Hddc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027168.2 | NP_081444.1 | 5 | 498 | ||
| GenBank mRNA | AK009957.1 | - | 5 | 439 | ||
| BC104360.1 | - | 5 | 498 | 40049707 | ||
| BC104361.1 | - | 5 | 498 | 40049709 | ||