siRNA Details
| Gene Symbol: Polr2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027231.1 | NP_081507.1 | 5 | 422 | ||
| GenBank mRNA | AF176516.2 | - | 1 | 1856 | ||
| AK007913.1 | - | 5 | 437 | |||
| AK165600.1 | - | 5 | 449 | |||
| AK169434.1 | - | 5 | 438 | |||
| BC024419.1 | - | 5 | 422 | 4983359 | ||

Polr2f
Genomic Map