siRNA Details
| Gene Symbol: Nipal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081205.1 | NP_001074674.1 | 6 | 1246 | ||
| GenBank mRNA | AK014427.1 | - | 6 | 1199 | ||
| AK035018.1 | - | 1 | 338 | |||
| AK075654.1 | - | 6 | 1250 | |||
| AK163026.1 | - | 1 | 533 | |||

Nipal1
Genomic Map