siRNA Details
| Gene Symbol: Rnf215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027859.2 | NP_082135.2 | 9 | 1673 | ||
| GenBank mRNA | AK002414.1 | - | 9 | 1631 | ||
| AK154140.1 | - | 7 | 1564 | |||
| BC103627.1 | - | 9 | 1597 | 6486672 | ||
| BM948157.1 | - | 1 | 263 | |||

Rnf215
Genomic Map