siRNA Details
| Gene Symbol: Mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028479.1 | NP_082755.1 | 4, 5 | 778 | ||
| GenBank mRNA | AK005551.1 | - | 3, 4 | 418 | ||
| AK217601.1 | - | 2, 3 | 195 | |||
| BC110993.1 | - | 3, 4 | 384 | 5363496 | ||
| BC116732.1 | - | 4, 5 | 778 | 40126051 | ||
| BC116734.1 | - | 4, 5 | 778 | 40126053 | ||

Mrgbp
Genomic Map