siRNA Details
Gene Symbol: 1700031F05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028496.1 | NP_082772.1 | 2, 3 | 291 | ||
GenBank mRNA | AK006578.1 | - | 2, 3 | 292 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700031F05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028496.1 | NP_082772.1 | 2, 3 | 291 | ||
GenBank mRNA | AK006578.1 | - | 2, 3 | 292 |