siRNA Details
| Gene Symbol: Acoxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028765.3 | NP_083041.1 | 7 | 853 | ||
| GenBank mRNA | AK004775.1 | - | 7 | 855 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acoxl| Gene Symbol: Acoxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028765.3 | NP_083041.1 | 7 | 853 | ||
| GenBank mRNA | AK004775.1 | - | 7 | 855 | ||