siRNA Details
| Gene Symbol: Gm10413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029288.3 | NP_083564.2 | 5 | 947 | ||
| GenBank mRNA | AK005583.1 | - | 5 | 937 | ||
| BC100412.1 | - | 5 | 947 | 30916569 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10413| Gene Symbol: Gm10413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029288.3 | NP_083564.2 | 5 | 947 | ||
| GenBank mRNA | AK005583.1 | - | 5 | 937 | ||
| BC100412.1 | - | 5 | 947 | 30916569 | ||