siRNA Details
| Gene Symbol: Bola3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175277.4 | NP_780486.1 | 3, 4 | 286 | ||
| GenBank mRNA | AK019037.1 | - | 2, 3 | 121 | ||
| AK028102.1 | - | 3, 4 | 287 | |||
| AK131950.1 | - | 295 | ||||
| AK160488.1 | - | 3, 4 | 290 | |||
| AK161878.1 | - | 58 | ||||
| AK218122.1 | - | 1, 2 | 103 | |||
| BC049748.1 | - | 3, 4 | 306 | |||
| BC116947.1 | - | 4, 5 | 324 | 40126266 | ||
| BC116949.1 | - | 4, 5 | 324 | 40126268 | ||

Bola3
Genomic Map