siRNA Details
| Gene Symbol: OR5I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006637.1 | NP_006628.1 | 1 | 442 | ||
| GenBank mRNA | BC069093.1 | - | 1 | 442 | 7216907 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5I1| Gene Symbol: OR5I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006637.1 | NP_006628.1 | 1 | 442 | ||
| GenBank mRNA | BC069093.1 | - | 1 | 442 | 7216907 | |