siRNA Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166221.1 | NP_001159693.1 | 10 | 1142 | ||
| NM_006649.3 | NP_006640.2 | 11 | 1298 | |||
| XM_017029237.1 | XP_016884726.1 | 13 | 1450 | |||
| XM_017029238.1 | XP_016884727.1 | 11 | 1480 | |||
| GenBank mRNA | AK094408.1 | - | 2 | 294 | ||
| AK291918.1 | - | 11 | 1265 | |||
| AK298578.1 | - | 10 | 1111 | |||
| AK304563.1 | - | 10 | 1163 | |||
| BC001149.1 | - | 11 | 1234 | 3355662 | ||
| BC009649.1 | - | 11 | 1236 | 3459192 | ||
| BC014987.1 | - | 11 | 1226 | 4901477 | ||

UTP14A
Genomic Map