siRNA Details
| Gene Symbol: CD300C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006678.4 | NP_006669.1 | 2 | 614 | ||
| XM_017024033.1 | XP_016879522.1 | 2 | 609 | |||
| XM_017024034.1 | XP_016879523.1 | 2 | 609 | |||
| GenBank mRNA | BC022279.1 | - | 2 | 586 | 4692025 | |
| X66171.1 | - | 2 | 464 | |||

CD300C
Genomic Map