siRNA Details
| Gene Symbol: WDR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006784.2 | NP_006775.1 | 3 | 333 | ||
| GenBank mRNA | AF083217.1 | - | 3 | 314 | ||
| AK292438.1 | - | 3 | 328 | |||
| BC058836.1 | - | 4 | 372 | |||
| BP281064.1 | - | 3 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR3| Gene Symbol: WDR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006784.2 | NP_006775.1 | 3 | 333 | ||
| GenBank mRNA | AF083217.1 | - | 3 | 314 | ||
| AK292438.1 | - | 3 | 328 | |||
| BC058836.1 | - | 4 | 372 | |||
| BP281064.1 | - | 3 | 333 | |||