siRNA Details
| Gene Symbol: OCLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022375.3 | NP_071770.1 | 1 | 530 | ||
| GenBank mRNA | AF142063.1 | - | 1 | 644 | ||
| BC069096.1 | - | 1 | 547 | 7216910 | ||
| BC093987.1 | - | 1 | 79 | 7939832 | ||
| BC093989.1 | - | 1 | 79 | 7939834 | ||

OCLM
Genomic Map