siRNA Details
| Gene Symbol: BRD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164326.1 | NP_001157798.1 | 17 | 2274 | ||
| NM_001300961.1 | NP_001287890.1 | 17 | 2270 | |||
| NM_001300962.1 | NP_001287891.1 | 16 | 2195 | |||
| NM_001300966.1 | NP_001287895.1 | 18 | 2362 | |||
| NM_006696.3 | NP_006687.3 | 18 | 2484 | |||
| NM_139199.1 | NP_631938.1 | 17 | 2265 | |||
| XM_005271855.2 | XP_005271912.1 | 18 | 2475 | |||
| XM_005271856.2 | XP_005271913.1 | 18 | 2475 | |||
| XM_005271857.2 | XP_005271914.1 | 17 | 2265 | |||
| XM_005271859.3 | XP_005271916.1 | 16 | 2046 | |||
| XM_005271860.3 | XP_005271917.1 | 18 | 2475 | |||
| XM_005271861.3 | XP_005271918.1 | 17 | 2256 | |||
| XM_011543110.1 | XP_011541412.1 | 18 | 2461 | |||
| XM_017008972.1 | XP_016864461.1 | 17 | 2265 | |||
| XM_017008973.1 | XP_016864462.1 | 17 | 2256 | |||
| XM_017008976.1 | XP_016864465.1 | 16 | 2046 | |||
| GenBank mRNA | AB209079.1 | - | 18 | 2430 | ||
| AF016270.1 | - | 17 | 2416 | |||
| AK289951.1 | - | 18 | 2444 | |||
| AK293621.1 | - | 16 | 2155 | |||
| AK297608.1 | - | 8 | 1553 | |||
| AK297756.1 | - | 17 | 2230 | |||
| AK312333.1 | - | 18 | 2444 | |||
| AL136823.1 | - | 17 | 2238 | |||
| BC008039.2 | - | 18 | 2426 | 3009915 | ||
| BC008076.2 | - | 18 | 2426 | 3009915 | ||
| X87613.1 | - | 17 | 2292 | |||
| X94234.1 | - | 16 | 2204 | |||

BRD8
Genomic Map