siRNA Details
| Gene Symbol: DBF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318060.1 | NP_001304989.1 | 8, 9 | 1203 | ||
| NM_001318061.1 | NP_001304990.1 | 9, 10 | 1218 | |||
| NM_001318062.1 | NP_001304991.1 | 9, 10 | 1249 | |||
| NM_006716.3 | NP_006707.1 | 9, 10 | 1310 | |||
| GenBank mRNA | AB028069.1 | - | 9, 10 | 1323 | ||
| AB028070.1 | - | 9, 10 | 1262 | |||
| AF160249.1 | - | 9, 10 | 1022 | |||
| AF160876.1 | - | 9, 10 | 1005 | |||
| AK292569.1 | - | 9, 10 | 1052 | |||
| AK302010.1 | - | 8, 9 | 832 | |||
| AK303175.1 | - | 9, 10 | 914 | |||
| BC022811.1 | - | 9, 10 | 907 | |||
| BC036045.1 | - | 9, 10 | 1116 | 5273059 | ||
| BC047693.1 | - | 9, 10 | 991 | 5753567 | ||

DBF4
Genomic Map