siRNA Details
| Gene Symbol: SPIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006717.2 | NP_006708.2 | 5 | 895 | ||
| GenBank mRNA | AF087864.1 | - | 5 | 775 | ||
| AF106682.1 | - | 5 | 776 | |||
| AF317228.2 | - | 4 | 740 | |||
| AK092017.1 | - | 8 | 1237 | |||
| AK289691.1 | - | 5 | 1080 | |||
| AK290009.1 | - | 5 | 763 | |||
| AK315854.1 | - | 6 | 1243 | |||
| AL136719.1 | - | 5 | 858 | |||
| BC013571.2 | - | 5 | 879 | |||
| BC114515.1 | - | 4 | 750 | 40031486 | ||
| BC114565.1 | - | 4 | 750 | 40031487 | ||
| BT007314.1 | - | 4 | 580 | |||
| CR533484.1 | - | 4 | 580 | |||

SPIN1
Genomic Map