siRNA Details
| Gene Symbol: GPR75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006794.3 | NP_006785.1 | 2 | 1142 | ||
| GenBank mRNA | AF072693.1 | - | 2 | 1037 | ||
| AK314885.1 | - | 2 | 1098 | |||
| BC067476.1 | - | 1 | 859 | 6971810 | ||
More Information
Set Membership: |
|

GPR75
Genomic Map