siRNA Details
Gene Symbol: C1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001733.4 | NP_001724.3 | 2 | 198 | ||
XM_017019915.1 | XP_016875404.1 | 2 | 272 | |||
GenBank mRNA | AK024951.1 | - | 1 | 898 | ||
AK222481.1 | - | 2 | 192 | |||
AK222491.1 | - | 2 | 192 | |||
AK291313.1 | - | 2 | 220 | |||
AK298445.1 | - | 2 | 213 | |||
AK303750.1 | - | 2 | 192 | |||
BC035220.1 | - | 2 | 198 | 4777940 | ||
M14058.1 | - | 2 | 202 | |||
X04701.1 | - | 2 | 190 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
