siRNA Details
| Gene Symbol: C1S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001734.3 | NP_001725.1 | 12 | 2291 | ||
| NM_201442.2 | NP_958850.1 | 12 | 2378 | |||
| XM_005253760.1 | XP_005253817.1 | 12 | 2270 | |||
| GenBank mRNA | AK025309.1 | - | 9 | 1766 | ||
| AK290295.1 | - | 12 | 2325 | |||
| AM886411.1 | - | 11 | 2013 | |||
| BC056903.1 | - | 12 | 2318 | 6199186 | ||
| J04080.1 | - | 12 | 2217 | |||
| M18767.1 | - | 12 | 2237 | |||

C1S
Genomic Map