siRNA Details
| Gene Symbol: TOMM34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006809.4 | NP_006800.2 | 2 | 358 | ||
| XM_005260254.2 | XP_005260311.1 | 2 | 291 | |||
| XM_011528501.1 | XP_011526803.1 | 2 | 177 | |||
| XM_017027600.1 | XP_016883089.1 | 2 | 405 | |||
| GenBank mRNA | AB085681.1 | - | 2 | 283 | ||
| AK000594.1 | - | 2 | 277 | |||
| AK129538.1 | - | 2 | 240 | |||
| AK222952.1 | - | 2 | 265 | |||
| AK302128.1 | - | 2 | 187 | |||
| AK307655.1 | - | 2 | 310 | |||
| BC001763.1 | - | 2 | 271 | 3544124 | ||
| BC007423.2 | - | 2 | 271 | 2960090 | ||
| BC014907.1 | - | 2 | 248 | 3922502 | ||
| BT020008.1 | - | 2 | 218 | |||
| BT020009.1 | - | 2 | 218 | |||
| CR456765.1 | - | 2 | 218 | |||
| U58970.1 | - | 2 | 249 | |||

TOMM34
Genomic Map