siRNA Details
| Gene Symbol: MLLT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006818.3 | NP_006809.1 | 2 | 1870 | ||
| GenBank mRNA | AK056089.1 | - | 2 | 1865 | ||
| AK057755.1 | - | 3 | 1103 | |||
| BC006471.2 | - | 1 | 992 | 2823316 | ||
| BC007079.1 | - | 2 | 1134 | |||
| BC008445.1 | - | 2 | 1134 | 4103485 | ||
| BC009624.1 | - | 2 | 1150 | 3878959 | ||
| BC021703.1 | - | 2 | 1208 | 4108071 | ||
| BC022448.1 | - | 2 | 2142 | 4815214 | ||
| BC032996.1 | - | 2 | 1167 | |||
| U16954.1 | - | 2 | 1342 | |||

MLLT11
Genomic Map