siRNA Details
| Gene Symbol: RAB40B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006822.2 | NP_006813.1 | 4 | 444 | ||
| XM_005256334.4 | XP_005256391.1 | 4 | 647 | |||
| XM_006722271.3 | XP_006722334.1 | 4 | 213 | |||
| XM_011523528.2 | XP_011521830.1 | 4 | 566 | |||
| XM_017024042.1 | XP_016879531.1 | 5 | 1079 | |||
| XM_017024043.1 | XP_016879532.1 | 4 | 556 | |||
| GenBank mRNA | AB209711.1 | - | 4 | 422 | ||
| AL047483.1 | - | 4 | 422 | |||
| BC018039.1 | - | 4 | 444 | 4801271 | ||
| U05227.1 | - | 4 | 363 | |||

RAB40B
Genomic Map