siRNA Details
Gene Symbol: ILVBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006844.4 | NP_006835.2 | 3 | 358 | ||
XM_005259717.4 | XP_005259774.1 | 3 | 505 | |||
XM_011527651.2 | XP_011525953.1 | 2 | 337 | |||
GenBank mRNA | AB209065.1 | - | 1 | 527 | ||
BC000109.1 | - | 2 | 302 | |||
BC011722.2 | - | 3 | 325 | 4300207 | ||
BC011761.2 | - | 3 | 251 | 3622453 | ||
BC126913.1 | - | 1 | 98 | |||
HY011076.1 | - | 3 | 358 | |||
U61263.1 | - | 3 | 233 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
