siRNA Details
| Gene Symbol: SLC27A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005094.3 | NP_005085.2 | 7 | 1252 | ||
| XM_017014222.1 | XP_016869711.1 | 8 | 1125 | |||
| GenBank mRNA | AF055899.1 | - | 6 | 967 | ||
| AK074831.1 | - | 4 | 300 | |||
| AK290222.1 | - | 7 | 1252 | |||
| AK314681.1 | - | 7 | 1092 | |||
| BC065003.1 | - | 7 | 1251 | 6023438 | ||

SLC27A4
Genomic Map