siRNA Details
| Gene Symbol: SLC27A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005094.3 | NP_005085.2 | 9 | 1543 | ||
| XM_017014222.1 | XP_016869711.1 | 10 | 1416 | |||
| GenBank mRNA | AF055899.1 | - | 8 | 1255 | ||
| AK027504.1 | - | 8 | 1123 | |||
| AK074831.1 | - | 6 | 591 | |||
| AK290222.1 | - | 9 | 1543 | |||
| AK314681.1 | - | 8 | 1563 | |||
| BC004268.1 | - | 2 | 103 | |||
| BC065003.1 | - | 9 | 1542 | 6023438 | ||

SLC27A4
Genomic Map