siRNA Details
| Gene Symbol: SNRNP27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006857.2 | NP_006848.1 | 3, 4 | 318 | ||
| NR_037862.1 | 3, 4 | 318 | ||||
| GenBank mRNA | AK291128.1 | - | 3, 4 | 299 | ||
| AK312041.1 | - | 3, 4 | 283 | |||
| BC017890.1 | - | 3, 4 | 303 | 4699198 | ||
| BU171817.1 | - | 3, 4 | 309 | |||
| X76302.1 | - | 3, 4 | 302 | |||

SNRNP27
Genomic Map