siRNA Details
| Gene Symbol: IFT27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177701.2 | NP_001171172.1 | 6, 7 | 885 | ||
| NM_006860.4 | NP_006851.1 | 6, 7 | 882 | |||
| XM_006724106.2 | XP_006724169.1 | 7, 8 | 646 | |||
| XM_017028540.1 | XP_016884029.1 | 6, 7 | 889 | |||
| GenBank mRNA | AL022729.1 | - | 6, 7 | 826 | ||
| BC000566.2 | - | 6, 7 | 863 | 3163341 | ||
| BT006815.1 | - | 6, 7 | 459 | |||
| CR456558.1 | - | 6, 7 | 501 | |||

IFT27
Genomic Map