siRNA Details
| Gene Symbol: TDRKH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083963.1 | NP_001077432.1 | 2 | 248 | ||
| NM_001083964.1 | NP_001077433.1 | 2 | 273 | |||
| NM_001083965.1 | NP_001077434.1 | 2 | 273 | |||
| NM_006862.3 | NP_006853.2 | 2 | 273 | |||
| XM_005244856.4 | XP_005244913.1 | 2 | 139 | |||
| XM_017000122.1 | XP_016855611.1 | 2 | 315 | |||
| XM_017000123.1 | XP_016855612.1 | 2 | 315 | |||
| XM_017000124.1 | XP_016855613.1 | 2 | 214 | |||
| XM_017000125.1 | XP_016855614.1 | 2 | 243 | |||
| XM_017000126.1 | XP_016855615.1 | 2 | 214 | |||
| XM_017000127.1 | XP_016855616.1 | 2 | 243 | |||
| XM_017000128.1 | XP_016855617.1 | 2 | 268 | |||
| XM_017000129.1 | XP_016855618.1 | 2 | 233 | |||
| XM_017000130.1 | XP_016855619.1 | 2 | 207 | |||
| GenBank mRNA | AF119121.2 | - | 2 | 261 | ||
| AF227192.1 | - | 2 | 261 | |||
| AK056402.1 | - | 2 | 208 | |||
| AK226092.1 | - | 2 | 186 | |||
| AK295948.1 | - | 2 | 144 | |||
| AK303063.1 | - | 2 | 169 | |||
| AK316279.1 | - | 2 | 224 | |||
| BC032690.1 | - | 2 | 180 | |||
| DA796638.1 | - | 2 | 273 | |||

TDRKH
Genomic Map