siRNA Details
| Gene Symbol: VAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112704.1 | NP_001106175.1 | 1 | 421 | ||
| NM_199131.2 | NP_954582.1 | 1 | 421 | |||
| GenBank mRNA | AK127095.1 | - | 1 | 418 | ||
| BC101694.1 | - | 1 | 205 | 8069200 | ||
| BC101696.1 | - | 1 | 205 | 8069202 | ||

VAX1
Genomic Map