siRNA Details
| Gene Symbol: LILRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130917.2 | NP_001124389.2 | 6 | 1232 | ||
| NM_001290270.1 | NP_001277199.1 | 5 | 1196 | |||
| NM_001290271.1 | NP_001277200.1 | 7 | 1671 | |||
| NM_006866.3 | NP_006857.2 | 7 | 1804 | |||
| XM_006722986.1 | XP_006723049.1 | 7 | 1804 | |||
| XM_011526385.1 | XP_011524687.1 | 7 | 1804 | |||
| XM_011526386.1 | XP_011524688.1 | 6 | 1768 | |||
| XM_011526387.1 | XP_011524689.1 | 6 | 1768 | |||
| XM_011526388.1 | XP_011524690.1 | 6 | 1768 | |||
| XM_011526389.1 | XP_011524691.1 | 5 | 1486 | |||
| XM_011526390.1 | XP_011524692.1 | 7 | 1769 | |||
| XM_011526391.1 | XP_011524693.1 | 6 | 1472 | |||
| XM_017026224.1 | XP_016881713.1 | 6 | 1507 | |||
| GenBank mRNA | AF025531.1 | - | 6 | 1223 | ||
| AK130170.1 | - | 6 | 1262 | |||
| AK222651.1 | - | 6 | 1228 | |||
| AK310088.1 | - | 7 | 1653 | |||
| BC017412.1 | - | 6 | 1234 | 4618777 | ||
| BC027916.1 | - | 6 | 1217 | 5223154 | ||
| CB046242.1 | - | 1 | 139 | |||
| EU915609.1 | - | 5 | 1165 | |||
| EU915610.1 | - | 6 | 1201 | |||
| U82275.1 | - | 6 | 1271 | |||
| U82276.1 | - | 5 | 1186 | |||
| U82277.1 | - | 4 | 395 | |||
| U82278.1 | - | 6 | 1184 | |||

LILRA2
Genomic Map