siRNA Details
| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198595.1 | NP_001185524.1 | 5 | 2368 | ||
| NM_006873.3 | NP_006864.2 | 4 | 2251 | |||
| GenBank mRNA | AK314464.1 | - | 4 | 2251 | ||
| AY026319.1 | - | 3 | 2156 | |||
| BC121064.2 | - | 3 | 2166 | 40118539 | ||

STON1
Genomic Map