siRNA Details
| Gene Symbol: GUSBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027386.2 | 6 | 931 | |||
| Gene Symbol: SMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X83300.1 | - | 4 | 672 | ||

GUSBP3
Genomic Map