siRNA Details
| Gene Symbol: PIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006875.3 | NP_006866.2 | 4 | 614 | ||
| GenBank mRNA | AK290931.1 | - | 4 | 502 | ||
| AK312745.1 | - | 4 | 502 | |||
| BC018111.1 | - | 4 | 485 | 3913552 | ||
| U77735.1 | - | 4 | 497 | |||

PIM2
Genomic Map