siRNA Details
| Gene Symbol: UPK1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281443.1 | NP_001268372.1 | 3 | 167 | ||
| NM_007000.3 | NP_008931.1 | 3 | 167 | |||
| XM_017026225.1 | XP_016881714.1 | 3 | 556 | |||
| GenBank mRNA | AF085807.1 | - | 2 | 135 | ||
| BC107097.1 | - | 2 | 140 | 40015698 | ||
| BC107098.1 | - | 2 | 140 | 40015700 | ||

UPK1A
Genomic Map