siRNA Details
| Gene Symbol: ADRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281437.1 | NP_001268366.1 | 9 | 1208 | ||
| NM_001281438.1 | NP_001268367.1 | 9 | 1212 | |||
| NM_007002.3 | NP_008933.2 | 10 | 1329 | |||
| NM_175573.2 | NP_783163.1 | 10 | 1325 | |||
| XM_005260257.1 | XP_005260314.1 | 11 | 1416 | |||
| XM_011528503.1 | XP_011526805.1 | 9 | 2061 | |||
| XM_017027602.1 | XP_016883091.1 | 10 | 2185 | |||
| XM_017027603.1 | XP_016883092.1 | 11 | 1437 | |||
| XM_017027604.1 | XP_016883093.1 | 10 | 2181 | |||
| XM_017027605.1 | XP_016883094.1 | 9 | 2082 | |||
| XM_017027606.1 | XP_016883095.1 | 9 | 2068 | |||
| XM_017027607.1 | XP_016883096.1 | 9 | 2064 | |||
| XM_017027608.1 | XP_016883097.1 | 8 | 1854 | |||
| XM_017027609.1 | XP_016883098.1 | 8 | 1833 | |||
| XM_017027610.1 | XP_016883099.1 | 7 | 1730 | |||
| GenBank mRNA | AK054985.1 | - | 2126 | |||
| AK307459.1 | - | 2347 | ||||
| BC010733.1 | - | 7 | 841 | |||
| BC017245.2 | - | 10 | 1225 | 4890786 | ||
| CR749213.1 | - | 2 | 5114 | |||
| D64154.1 | - | 10 | 1220 | |||

ADRM1
Genomic Map