siRNA Details
| Gene Symbol: Avp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016992.2 | NP_058688.2 | 3 | 560 | ||
| GenBank mRNA | M25646.1 | - | 3 | 560 | ||
| M64785.1 | - | 908 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Avp| Gene Symbol: Avp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016992.2 | NP_058688.2 | 3 | 560 | ||
| GenBank mRNA | M25646.1 | - | 3 | 560 | ||
| M64785.1 | - | 908 | ||||