siRNA Details
| Gene Symbol: Jund | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286966.1 | NP_001273895.1 | 1 | 1360 | ||
| NM_138875.4 | NP_620230.1 | 1 | 1360 | |||
| GenBank mRNA | BC062053.1 | - | 1 | 1360 | 5598945 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jund| Gene Symbol: Jund | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286966.1 | NP_001273895.1 | 1 | 1360 | ||
| NM_138875.4 | NP_620230.1 | 1 | 1360 | |||
| GenBank mRNA | BC062053.1 | - | 1 | 1360 | 5598945 | |