siRNA Details
| Gene Symbol: Npy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012614.2 | NP_036746.1 | 4 | 522 | ||
| GenBank mRNA | JX157883.1 | - | 4 | 343 | ||
| M15880.1 | - | 4 | 498 | |||
| M20373.1 | - | 4 | 477 | |||
| Gene Symbol: LOC100912228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749745.3 | XP_003749793.1 | 535 | |||

Npy
Genomic Map